missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygoprep™ 60 to 30

Brand:  Macherey-Nagel™ 711250.1000

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10775922

  • £177.00 / 1000g
Estimated Shipment: 16-08-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

25 to 40 μm
POLYGOPREP 60 to 30
2 to 8
POLYGOPREP
Polygoprep
Liquid Chromatography
60 Å
Liquid Chromatography
1 kg
350 m2/g
Silica
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers