missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygosil™ Phase

Brand:  Macherey-Nagel™ 711020.10

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10134741

  • £32.45 / 10g
Estimated Shipment: 16-08-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

10 μm
Liquid Chromatography
Polygosil Phases
60 Å
10 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers