missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Polygosil™ Phase

Brand:  Macherey-Nagel™ 711600.10

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10378481

  • £65.85 / 10g
Estimated Shipment: 16-08-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

7 μm
Liquid Chromatography
Polygosil Phases
300 Å
10 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers