missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyphosphoric Acid, 105% H3PO4 Basis, Honeywell Fluka™
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
pH | Acidic |
Color | Colorless |
Melting Point | 16°C |
Boiling Point | 300°C |
Density | 2.06g/cm3 at 20°C |
Odor | Odorless |
Packaging | Poly Bottle |
Quantity | 100 mL |
Linear Formula | Hn+2PnO3n+1 |
Physical Form | Liquid |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'