missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyphosphoric acid, pure, >84% phosphate (as P2O5), Thermo Scientific Chemicals
£60.60 - £352.59
Chemical Identifiers
CAS | 8017-16-1 |
---|---|
Molecular Formula | (HO3P)n |
MDL Number | MFCD00084480 |
InChI Key | NBIIXXVUZAFLBC-UHFFFAOYSA-N |
IUPAC Name | Polyphosphoric acid |
SMILES | [H]OP(O)(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10542411
|
Thermo Scientific Acros
196950010 |
1 kg | Plastic bottle |
£60.60
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10481431
|
Thermo Scientific Acros
196950025 |
2.5 kg | Plastic bottle |
£125.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11924101
|
Thermo Scientific Acros
196950100 |
10 kg | Plastic drum | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
8017-16-1 | |
MFCD00084480 | |
Polyphosphoric acid |
(HO3P)n | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
[H]OP(O)(O)=O |
Specifications
8017-16-1 | |
2.0500g/mL | |
(HO3P)n | |
MFCD00084480 | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
Polyphosphoric acid | |
Pure |
550.0°C | |
2.05 | |
Viscous Liquid | |
Solubility in water: may decompose. Other solubilities: insoluble in hydrocarbons and halogenated hydrocar, soluble in alcohols | |
[H]OP(O)(O)=O | |
≥82% (as P2O5) (Alkalimetry) | |
Polyphosphoric acid |
Safety and Handling
Recommended Storage : May darken over time
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Polyphosphoric acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only