missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Polypropylene 12 Channel Reagent Reservoir

Polypropylene 12 Channel Reagent Reservoir’s V-shaped wells minimize waste by allowing complete sample withdrawal

Brand:  Bel-Art™ SP Scienceware™ F37880-1200

36.49 GBP valid until 2025-04-18
Use promo code "24095" to get your promotional price.


Product Code. 15565604

  • £64.25 / Pack of 8
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Each channel holds 5ml; numbered to facilitate serial dilution
  • Chemical resistant polypropylene construction; protective cover included
  • Can be used with single, 8 or 12 channel pipettors
  • Wide profile provides added stability; measures 12.7L x 5.8cm W (5 x 2.25 in.)
  • Individually sealed and sterilized
  • Autoclavable at 121°C (250°F)
Specifications

Specifications

White
12.7 x 5.8 x 3.2cm (5 x 2.25 x 1.25in.)
Sterile
Autoclavable
60 mL
Polypropylene
Reagent Reservoir
Flat
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Use code 24095 to receive this promotion.
    Promo Code: 24095

    Expires2025-04-15

    Days

    26

    Hours

    2

    Minutes

    6

    Seconds

    58
    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 17/03/2025 and 15/04/2025 subject to availability of the product in stock. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy or if the product is no longer available in stock. Other restrictions may apply.

    Terms & Conditions Learn More
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ SP Scienceware™ Polypropylene 12 Channel Reagent Reservoir > Dimensions (L x W x H): 12.7 x 5.8 x 3.2cm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.