Learn More
Fisherbrand™ Pasteur-plast Pipets
Use these pipettes for simple and safe liquid handling. Fisherbrand™ Pasteur-plast Pipets are inert to biological fluids and most acids. The low affinity of the surface reduces the loss of cells and valuable proteins through binding.
Brand:  Fisherbrand™ FB55343
Product Code. 11577722
  • £21.32 / Pack of 500
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Cross-contamination and carry-over simply cannot occur so that tedious clean-ups are unnecessary. In addition, there is no danger of glass breakage with the resulting danger of infectious material escaping.
  • New soft, elastic bulb
  • Unbreakable – thus reduced risk of glass injury
  • Burr-free orifice for neat, clean and precise dispensing
  • Embossed, external graduations
  • BSE-free can be used for food
Specifications
Specifications
Polypropylene
Food analysis
500 Pack
150 mm
Non-sterile
Capillary aspiration tip
1 mL
Pasteur Pipet
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Pasteur-plast Pipets >