Learn More
Hirschmann™ Polypropylene Thread Adapter
Product Code. 10195260
Click to view available options
Thread Size:
25 mm
28 mm
32 mm
38 mm
38/430 mm
40 mm
A 32 to A 38
A 32 to A 45
A 45 to S 40
Unit Size:
Each
Product Code. 10195260
Brand: Hirschmann™ 9313502
Only null left
Add to basket
Add to basket
Polypropylene (PP) Threaded adapter for adaptation on the most common bottles.
  • For the adaption on the most common bottles
Specifications
Product Type Adapter
Material Polypropylene
For Use With (Equipment) ceramus™ classic and EM-dispenser
Quantity 1 Ea.
Thread Size 25 mm
Diameter (Metric) Neck 32 mm