missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Polypropylene Barbed Y-Connector

Ideal Y-connector for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, and food processing. Cole-Parmer™ Polypropylene Barbed Y-Connector

Brand:  Masterflex™ 06295-50

Product Code. 12419590

This item is not returnable. View return policy

Description

Description

  • Autoclavable
  • Durable polypropylene
  • Barbed for secure fit
  • Clear
Specifications

Specifications

Autoclavable
Polypropylene
Fits 12mm I.D. Tubing
47 mm
Barbed Y-Connector
9 mm
74 mm
20 Pack
Clear
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Masterflex™ Polypropylene Barbed Y-Connector > For use with: 12mm I.D. Tubing

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.