Learn More
Buerkle™ Polypropylene Barrel Screw Joint
Product Code. 10126370 Shop All Bürkle Products
Click to view available options
Size:
Mauser 2 in., Coarse Outer Thread
R 2 in., Fine Steel Outer Thread
Tri-Sure, Coarse Outer Thread
Unit Size:
Each
Product Code. 10126370
Brand: Buerkle™ 56003180
Only null left
Add to basket
Add to basket
Buerkle™ Polypropylene Barrel Screw Joint for secure connection in the barrel in three different thread models.
  • Barrel screw joint made of PP for secure connection in the barrel in three different thread models
  • For drawing off liquids that are not flammable
  • The container connectors make decanting even easier and safer with their more secure connection to barrels
  • The connectors secure the pump in the barrel, enable a vertical position change and allow the required air exchange
TRUSTED_SUSTAINABILITY
Specifications
For Use With PTFE Barrel Pump
Material Polypropylene
Size Tri-Sure, Coarse Outer Thread
Type Barrel Screw Joint