missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kautex™ Polypropylene Blister Closures
Click to view available options
Color:
Black
Red
Includes:
With Tamper Evident Ring
With Tamper Evident Ring Polypropylene/Red, With Venting System and Perforated PTFE Seal
Description
- Tamper evident rings
- PTFE cup seal or perforated seal
- Available with venting system
- Available in black or red to differentiate available features
Specifications
Color | Black |
Includes | With Tamper Evident Ring |
Material | Polypropylene |
Product Type | Blister Closure |
For Use With (Equipment) | 500 to 2500 mL Bottle |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'