Learn More
Fisherbrand™ Polypropylene Blue Pipet Tip
Make pipetting simple and safer for even the most sensitive applications. Fisherbrand™ Polypropylene Blue Pipet Tips are perfectly fit for manual or electronic, single or multichannel pipets.
Brand: Fisherbrand™
23.16 GBP valid until 2024-12-31
Use promo code "21921" to get your promotional price.
Code : 41
Additional Details : Weight : 0.67500kg
Description
- Made of high quality polypropylene
- Tight fit due to the precisely fitting cone
- Optimal handling due to the stable shape
- Autoclavable
- Guaranteed dust-free due to special ionization treatment
- Available pre-sterilized or non-sterile
- Excellent transparency due to high quality surfaces
- Minimal residual wetting due to controlled quality of raw materials
- Excellent volume accuracy due to precise tip orifice
- Practice-oriented tip volumes
- Aeroject™ tips with special filters made of hydrophobic PE-HD as
- Aerosol barrier and for protection against contamination
- Suitable for all commonly used pipet types
Specifications
200 to 1000 μL | |
Blue | |
Autoclavable | |
Polypropylene |