missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ratiolab™ Polypropylene Blue Pipet Tip E
Use these pipet tips for simple and safe liquid handling in the most sensitive applications. Ratiolab™ Polypropylene Blue Pipet Tips E are ideal for manual or electronic, single or multichannel pipets.
£10.95 - £35.85
Specifications
Volume (Metric) | 100 to 1000 μL |
---|---|
Color | Blue |
Autoclavable | Autoclavable |
Length (Metric) | 6.9cm |
Certifications/Compliance | CE, IVD |
Product Code | Brand | Volume (Metric) | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Format | Price | Quantity & Availability | ||||
10789521
|
Ratiolab™
2100610 |
100 to 1000 μL | Bulk |
£26.30
Pack of 1000 |
Estimated Shipment: 20-12-2024
Log in to see stock. |
||||
10169972
|
Ratiolab™
2130612 |
100 to 1000 μL | Rack |
£35.85
Pack of 960 |
Estimated Shipment: 16-12-2024
Log in to see stock. |
||||
11836943
|
Ratiolab™
2130612 |
100 to 1000 μL | Rack |
£10.95
Pack of 120 |
In Stock
Log in to see stock. |
||||
Description
- Ideal for all commercial pipets
- Made of high quality polypropylene
- Tight fit due to the precisely fitting cone
- Optimal handling due to stable shape
- Autoclavable
- Guaranteed dust free due to special ionization treatment
- Available in pre-sterilized or non-sterile
- Excellentr4 transparency and high quality surfaces
- Minimal residual wetting
- Excellent volume accuracy due to precise tip orifice
- Practice-oriented tip volumes
Specifications
100 to 1000 μL | |
Autoclavable | |
CE, IVD | |
Autoclavable | |
Polypropylene |
Blue | |
6.9cm | |
Ideal for All Commercially Available Pippets | |
Universal Fit |