missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Polypropylene Cable Ties

Product Code. p-8006910 Shop All Cole-Parmer Products
Click to view available options
Color:
Black
White
Length (Metric):
10 cm
20 cm
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 11723391

Brand: Cole-Parmer™ 0683085

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Keep items bundled during autoclaving with these ties. Cole-Parmer™ Polypropylene Cable Ties are ideal for tubing bundles.

  • Polypropylene (PP) cable ties resist a number of chemicals
  • UV-resistant
  • Sold in packs of 1000

Specifications

Type Cable Tie
Material Polypropylene
Color White
Length (Metric) 10 cm
Quantity 1000/Pk.
For Use With (Application) Ideal for tubing bundles
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.