missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Polypropylene, Carboy
Click to view available options
Capacity (Metric):
40 L
Description
- Interchangeable caps and adapters provide top connection flexibility
- Rectangular ergonomic design
- Constructed from autoclavable USP Class VI, FDA grade materials
Specifications
Accuracy | ±5% |
Autoclavable | Autoclavable |
Capacity (English) | 10.6 gal. |
Capacity (Metric) | 40 L |
Closure Material | Polypropylene |
Closure Size | 120 mm |
Color | White |
Dimensions (L x W x H) | 434 x 249 x 772 mm |
For Use With (Application) | For storing and dispensing solvents, buffers, reagents and powders. |
Height (English) | 30.4 in. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'