missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Polypropylene Clear Autoclave Bags
Designed to be strong, pliable, lean and puncture resistant. Fisherbrand™ Polypropylene Clear Autoclave Bags withstand high temperatures for autoclaving.
Brand: Fisherbrand™ 11593342
Description
Designed to be strong, pliable, lean and puncture resistant. Fisherbrand™ Polypropylene Clear Autoclave Bags withstand high temperatures for autoclaving.
- For high temperature sterilisation (134°C) to a maximum of 140°C
- 40μm thickness
- High temperature polypropylene bags for the decontamination and inactivation of particularly resistant biological waste
- For safe disposal of biohazard waste
- Specifically designed for contaminated waste disposal in autoclaves or incinerators
- Printed with a biohazard warning
Specifications
Plain translucent bag, no print | |
Clear | |
40 μm | |
620 x 780 mm | |
620 mm |
Autoclavable | |
Polypropylene | |
100/Pk. | |
134°C to 140°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction