missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Clear Autoclave Bags

Designed to be strong, pliable, lean and puncture resistant. Fisherbrand™ Polypropylene Clear Autoclave Bags withstand high temperatures for autoclaving.

Brand:  Fisherbrand™ 11600312

 View more versions of this product

Product Code. 16247971

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Description

Description

Designed to be strong, pliable, lean and puncture resistant. Fisherbrand™ Polypropylene Clear Autoclave Bags withstand high temperatures for autoclaving.

  • For high temperature sterilisation (134°C) to a maximum of 140°C
  • 40μm thickness
  • High temperature polypropylene bags for the decontamination and inactivation of particularly resistant biological waste
  • For safe disposal of biohazard waste
  • Specifically designed for contaminated waste disposal in autoclaves or incinerators
  • Printed with a biohazard warning
Specifications

Specifications

Printed with biohazard waste identification
Clear with Biohazard Warning
40 μm
310 x 660 mm
310 mm
Autoclavable
Polypropylene
100/Pk.
134°C to 140°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Polypropylene Clear Autoclave Bags > 310W x 660mmD

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.