missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Polypropylene Reducing Hose Connectors

Product Code. 10739792 Shop All Bürkle Products
Click to view available options
For Use With:
4 to 8/12 to 16 mm Hose
4 to 8/8 to 12 mm Hose
8 to 12/12 to 16 mm Hose
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 10739792

Brand: Buerkle™ 87030812

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use Buerkle™ Polypropylene Reducing Hose Connectors to make a connections between larger and smaller hoses.

  • Polypropylene
  • Sterilizable
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EU Foodstuffs, RoHS
Autoclavable Autoclavable
Material Polypropylene
For Use With 8 to 12/12 to 16 mm Hose
Packaging Quantity 10/Pk.; 10/Scale
Color White
Product Type Reducing Hose Connector
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.