Learn More
Polypropylene Disposal Bins
Click to view available options
Capacity (Metric):
7 L
Unit Size:
Pack of 40
Description
Ideal for cytotoxic and cytostatic waste which, to meet regulations, should be disposed of separately from other healthcare waste. The SHARPSGUARD™ cyto lid assists safe, easy collection of items including giving sets, syringes and gloves. Color-coded with a purple lid, handle and label, it can be safely transported for incineration after closure. Absorbent pads are included. The container is made to BS EN ISO 23907.
Specifications
Specifications
| Capacity (Metric) | 7 L |