missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ EcoSpatula Single-Use Biodegradable Spatula

Environmentally friendly, biodegradable, single-use spatula. Cole-Parmer™ EcoSpatula Single-Use Biodegradable Spatula is resistant to dilute acids and bases.

Brand:  Cole-Parmer™ 17261

Product Code. 15352291

  • £60.15 / Pack of 300
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Reduces landfill waste
  • Biodegrades an average of 13.9% in the first 162 days
  • Polypropylene construction with EcoPure additive to promote biodegredation
  • FDA compliant for food contact applications
  • Resistant to dilute acids and bases
  • Shatter-resistant in liquid nitrogen
  • Suitable for compounding of sterile applications
  • Features both a scoop and spoon end with a hollow core to reduce heat and cold transfer
  • Autoclavable
Specifications

Specifications

300 Pk.
Autoclavable
Polypropylene
144 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers