Learn More
Masterflex™ Polypropylene Female Luer Bulkhead Fitting
Brand:  Masterflex™ 45508-34
 View more versions of this product
Product Code. 15159014
  • £134.00 / Pack of 25
Estimated Shipment: 18-06-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Female Luer Bulkhead Fitting
Fits 1/4-28 UNF thread to 1/8 in. hose barb
25 Pack
Polypropylene
Ideal for a variety of applications, including syringes or small tubing