missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Polypropylene Graduated Microtubes
GREENER_CHOICE

Product Code. 10518721 Shop All Eppendorf Products
Click to view available options
Color:
Amber
Assorted
Blue
Clear
Green
Red
Yellow
Capacity (Metric):
0.5 mL
1.5 mL
2 mL
Purity or Quality Grade:
Biopur
Biopur™
Eppendorf Quality
PCR Clean
Unit Size:
Pack of 100
Pack of 1000
Pack of 50
Pack of 500

Product Code. 10518721

Brand: Eppendorf™ 0030121589

Only null left
Add to basket
Add to basket

Use these tubes for safe handling of valuable and toxic samples as well as radioactive substances. Eppendorf™ Polypropylene Graduated Microtubes feature a hinged lid with a small hook that clips around the rim of the tube.

Use these tubes for safe handling of valuable and toxic samples as well as radioactive substances. EppendorfTM Polypropylene Graduated Microtubes feature a hinged lid with a small hook that clips around the rim of the tube.

  • Biohazardous substances can be removed, free of aerosols, through a thin membrane in the center of the lid with a cannula
  • Lid easily opens with one hand
  • These tubes for 0.5mL, 1.5mL and 2mL have a graduation scale and writing surfaces
  • The 0.5mL tube is optimized for use in PCR* experiments
  • Assorted color packs are a mixture of yellow, red, blue, green and colorless
  • Autoclavable micropestle for re-suspending pellets
  • Can be centrifuged at up to 23,500xgr
  • Purity grade: Biopur™
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Color Clear
Material Polypropylene
For Use With (Equipment) PCR, Centrifugation
No. per Pack 100Pack
Sterility Sterile
Purity or Quality Grade Biopur™
Graduated Yes
Closure Type Hinged Snap Cap
Max. RCF 30,000 x g
Afficher plus Afficher moins
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis