missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Polypropylene Graduated Microtubes
GREENER_CHOICE

Product Code. 10767411 Shop All Eppendorf Products
Click to view available options
Color:
Amber
Assorted
Blue
Clear
Green
Red
Yellow
Capacity (Metric):
0.5 mL
1.5 mL
2 mL
Purity or Quality Grade:
Biopur
Biopur™
Eppendorf Quality
PCR Clean
Unit Size:
Pack of 100
Pack of 1000
Pack of 50
Pack of 500
This item is not returnable. View return policy

Product Code. 10767411

Brand: Eppendorf™ 0030120175

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use these tubes for safe handling of valuable and toxic samples as well as radioactive substances. Eppendorf™ Polypropylene Graduated Microtubes feature a hinged lid with a small hook that clips around the rim of the tube.

Use these tubes for safe handling of valuable and toxic samples as well as radioactive substances. EppendorfTM Polypropylene Graduated Microtubes feature a hinged lid with a small hook that clips around the rim of the tube.

  • Biohazardous substances can be removed, free of aerosols, through a thin membrane in the center of the lid with a cannula
  • Lid easily opens with one hand
  • These tubes for 0.5mL, 1.5mL and 2mL have a graduation scale and writing surfaces
  • The 0.5mL tube is optimized for use in PCR* experiments
  • Assorted color packs are a mixture of yellow, red, blue, green and colorless
  • Autoclavable micropestle for re-suspending pellets
  • Can be centrifuged at up to 23,500xgr
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Color Blue
Material Polypropylene
For Use With (Equipment) PCR, Centrifugation
No. per Pack 1000/Pk.
Purity or Quality Grade Eppendorf Quality
Graduated Yes
Closure Type Hinged Snap Cap
Max. RCF 30,000 x g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.