missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Polypropylene Graduated Microtubes
Use these tubes for safe handling of valuable and toxic samples as well as radioactive substances. Eppendorf™ Polypropylene Graduated Microtubes feature a hinged lid with a small hook that clips around the rim of the tube.
£32.18 - £93.52
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Polypropylene |
Graduated | Yes |
Closure Type | Hinged Snap Cap |
Product Code | Brand | Color | Capacity (Metric) | Purity or Quality Grade | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Capacity (Metric) | Purity or Quality Grade | Price | Quantity & Availability | |||
10458312
|
Eppendorf™
0030121155 |
Amber | 0.5 mL | PCR Clean |
£67.65
Pack of 500 Qty Price Break Available |
In Stock
Log in to see stock. |
|||
10160431
|
Eppendorf™
0030120191 |
Amber | 1.5 mL | PCR Clean |
£72.14
Pack of 1000 |
In Stock
Log in to see stock. |
|||
10252851
|
Eppendorf™
0030120248 |
Amber | 2 mL | PCR Clean |
£93.52
Pack of 1000 |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||
10548841
|
Eppendorf™
0030121694 |
Assorted | 1.5 mL | Eppendorf Quality |
£50.60
Pack of 1000 |
In Stock
Log in to see stock. |
|||
10702132
|
Eppendorf™
0030121686 |
Assorted | 2 mL | Eppendorf Quality |
£69.73
Pack of 1000 |
Estimated Shipment: 16-12-2024
Log in to see stock. |
|||
10622731
|
Eppendorf™
0030121708 |
Assorted | 0.5 mL | Eppendorf Quality |
£50.60
Pack of 500 |
In Stock
Log in to see stock. |
|||
10767411
|
Eppendorf™
0030120175 |
Blue | 1.5 mL | Eppendorf Quality |
£66.21
Pack of 1000 |
In Stock
Log in to see stock. |
|||
10393431
|
Eppendorf™
0030121570 |
Clear | 0.5 mL | Biopur |
£32.18
Pack of 50 |
In Stock
Log in to see stock. |
|||
10518721
|
Eppendorf™
0030121589 |
Clear | 1.5 mL | Biopur™ |
£32.83
Pack of 100 |
In Stock
Log in to see stock. |
|||
10241931
|
Eppendorf™
0030121597 |
Clear | 2 mL | Biopur |
£43.67
Pack of 100 |
In Stock
Log in to see stock. |
|||
10292611
|
Eppendorf™
0030121023 |
Clear | 0.5 mL | Eppendorf Quality |
£46.74
Pack of 500 |
In Stock
Log in to see stock. |
|||
10038760
|
Eppendorf™
0030120094 |
Clear | 2 mL | Eppendorf Quality |
£64.56
Pack of 1000 |
In Stock
Log in to see stock. |
|||
10509691
|
Eppendorf™
0030120086 |
Clear | 1.5 mL | Eppendorf Quality |
£47.24
Pack of 1000 |
In Stock
Log in to see stock. |
|||
10170381
|
Eppendorf™
0030120183 |
Green | 1.5 mL | Eppendorf Quality |
£66.21
Pack of 1000 |
In Stock
Log in to see stock. |
|||
10282851
|
Eppendorf™
0030120167 |
Red | 1.5 mL | Eppendorf Quality | N/A | ||||
10708571
|
Eppendorf™
0030120159 |
Yellow | 1.5 mL |
£66.21
Pack of 1000 |
Estimated Shipment: 16-12-2024
Log in to see stock. |
||||
Description
Use these tubes for safe handling of valuable and toxic samples as well as radioactive substances. EppendorfTM Polypropylene Graduated Microtubes feature a hinged lid with a small hook that clips around the rim of the tube.
- Biohazardous substances can be removed, free of aerosols, through a thin membrane in the center of the lid with a cannula
- Lid easily opens with one hand
- These tubes for 0.5mL, 1.5mL and 2mL have a graduation scale and writing surfaces
- The 0.5mL tube is optimized for use in PCR* experiments
- Assorted color packs are a mixture of yellow, red, blue, green and colorless
- Autoclavable micropestle for re-suspending pellets
- Can be centrifuged at up to 23,500xgr
Specifications
Autoclavable | |
Polypropylene | |
Yes | |
Hinged Snap Cap |