missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Polypropylene Labels for BradyJet J4000
This B-7425J polypropylene material is ideal for GHS and other full-color identification applications where resistance to harsh chemicals is required.
£264.00 - £682.00
Specifications
Autoclavable | Autoclavable |
---|---|
Adhesive Type | Permanent Acrylic |
Color | White |
Finish | Matte |
Material | Polypropylene (Inkjet-receptive) |
Product Code | Brand | Height (Metric) | Width (Metric) | Vertical Repeat (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Width (Metric) | Vertical Repeat (Metric) | Price | Quantity & Availability | |||
17499212
|
Brady™
152778 |
85 mm | 55 mm | 92.1 mm |
£274.00
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||
17429222
|
Brady™
152781 |
133.4 mm | 203.2 mm | 142.9 mm |
£342.00
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||
17409222
|
Brady™
152779 |
155 mm | 104.9 mm | 161.9 mm |
£418.00
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||
17419222
|
Brady™
152780 |
254 mm | 177.8 mm | 266.7 mm |
£264.00
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||
17439222
|
Brady™
152782 |
279.4 mm | 203.2 mm | 292.1 mm |
£682.00
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||
Description
- Matte white polypropylene with acrylic adhesive is Inkjet printable
- Low-cost, full-color GHS labeling option withstands chemical exposure
- Service temperature of -80°C to 70°C
- An ink cartridge is required to print this material
Specifications
Autoclavable | |
White | |
Polypropylene (Inkjet-receptive) | |
Non-sterile | |
BradyJet J4000 Color Label Printer | |
Rectangle | |
0.13 mm |
Permanent Acrylic | |
Matte | |
Ink Jet | |
Hazard Communication Identification, Equipment Labeling, Laboratory Labeling, General Identification | |
1 | |
-80°C to +70°C |