Learn More
Masterflex™ Polypropylene Luer Fitting
Click to view available options
For Use With:
Female Luer x 1/4 to 28 in. UNF
Female Luer x Female Luer, Elbow
Female x Female x Female
Male LL to 1/4 to 28 UNF Thread
Male LL to 10 to 32 Thread
Male x Female x Female
Unit Size:
Pack of 25
Description
- Three barb styles: standard, wide-bore, and low-profile.
- Wide-bore barbs offer enhanced fluid flow through the fitting and are usable with any flexible tubing.
- Low-profile barbs are ideal for use with semi-rigid tubing, such as nylon, polyethylene, or polypropylene.
- Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable.
- All parts meet tight dimensional tolerances for increased reliability and process purity.
- Choose from a range of sizes, 1/16in. to 1/4in., with a variety of connection options.
- Pack of 25
Specifications
Specifications
| Certifications/Compliance | ISO 594-1 standards |
| Material | Polypropylene |
| For Use With | Male LL to 1/4 to 28 UNF Thread |
| Packaging Quantity | 25 Pack |
| Product Type | Luer Fitting |