missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Polypropylene ADCF Luer Fittings

Product Code. 15122187
Click to view available options
For Use With:
4 in. J Barb Adapter
4 in. L Barb Adapter
Male LL x 1/4 in. Hose Barb
Male LL x 1/8 in. Hose Barb
Male Luer to 1/4 in. Barb Adapter
Male Luer x 1/8 in. Wide Bore Hose Barb
Male luer lock plug
Unit Size:
Pack of 25
This item is not returnable. View return policy

Product Code. 15122187

Brand: Masterflex™ 3080018

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ensure process integrity with full material traceability. Cole-Parmer™ Polypropylene ADCF Luer Fittings are ideal for a variety of critical applications, including syringes or small tubing. These fittings are inspected to be free from contaminants, and are fully traceable.

Ensure process integrity with full material traceability. Cole-ParmerTM Polypropylene ADCF Luer Fittings are ideal for a variety of critical applications, including syringes or small tubing. These fittings are inspected to be free from contaminants, and are fully traceable.

All parts meet tight dimensional tolerances for increased reliability and process purity. Choose from a range of sizes, 1⁄16" to 1⁄4", with a variety of connection options.
  • Manufactured from 100% virgin raw materials in a Class 100,000 environment
  • In compliance with ISO 594-1 standards
  • Manufactured of animal-free natural polypropylene
  • Meets USP Class VI specifications
  • Very good chemical resistance
  • Max temperature 190°F

Specifications

Material Polypropylene
For Use With Male Luer x 1/8 in. Wide Bore Hose Barb
For Use With (Application) Ideal for a variety of critical applications, including syringes or small tubing.
Packaging Quantity 25 Pack
Product Type Polypropylene ADCF Luer Fitting
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.