missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Polypropylene Measuring Pipettes

High clarity polypropylene construction. Calibrated to deliver and resistant to breakage.

£111.00 - £218.00

Specifications

Autoclavable Not Autoclavable
Calibrated Yes
Calibration To Deliver
Material Polypropylene
Graduated Yes
View More Specs

Products 3
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
11813920
View Documents
BRAND™
27613
10 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11718168
View Documents
BRAND™
27607
1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11803920
View Documents
BRAND™
27612
5 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Polypropylene construction, type 3 plastic
  • Calibrated to deliver (TD, Ex)
  • Subdivisions: 0.1mL
  • Supplied in packs of 12
  • Exposure to temperatures up to 60°C will not cause permanent exceeding of error limits
  • Cleaning temperature below 60°C is recommended to preserve marks and inscriptions
Specifications

Specifications

Not Autoclavable
To Deliver
Yes
0.1 mL
12/Pk.
Yes
Polypropylene
Blue
Printed
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.