missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Polypropylene Micro Tubes Cap Insert
Polypropylene Micro Tubes Cap Insert for use with all non-tamper evident screw caps allows for easy identification.
Brand: BRAND™ 780722
18.75 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- For micro tubes with attached or bulk screw caps
- Suitable for use with non-tamper evident caps
Specifications
Microtube Cap Insert | |
Red | |
Microtubes, Screw Caps |
Polypropylene | |
-196°C to +121°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction