Learn More
Brand™ Polypropylene Microcentrifuge Tube
Brand™ Microcentrifuge Tubes with Lid fit most common rotors and may be spun up to 30,000 RCF.
Brand: Brand™ 780507
43.80 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Free from flash, air bubbles and streaks
- Uniform wall thickness for consistent, repeatable results
- Caps for a tight, leak-free seal that is easy to open, even when wearing gloves
- Simplify sample identification
Specifications
Locking Lid | |
10,000 x g | |
Clear | |
1000/Pk. |