Learn More
Kartell™ Polypropylene Micropipet Tips
Designed with high clarity and excellent water-repellent properties to keep sample retention within the tip to a minimum. Kartell™ Polypropylene Micropipet Tips feature precise centering of the orifice at the end of the tip to guarantee accurate delivery of the sample.
Brand:  Kartell™ 0098400
34.66 GBP valid until 2024-12-31
Use promo code "21921" to get your promotional price.
 View more versions of this product
Product Code. 12024628
  • £41.40 / Pack of 250
Estimated Shipment: 18-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Cadmium-free polypropylene construction provides high clarity and water-repellent properties
  • Specially designed for Kartell automatic pipets, but deliver excellent results with most available micropipet brands 
  • Autoclavable to 121°C for 20 minutes
  • Precision delivery of sample, minimal sample retention with tip 
  • Packaging selection satisfies a variety of needs:
    • bulk bag
    • packed in racks
    • refills for reloading racks
Specifications
Specifications
1000 to 5000 μL
Autoclavable
Bulk
Gilson™ Pipetman™, Nichiryo™, Kartell Pipette, Socorex™ Pipette
Light Blue
CE, IVD
Low Retention
Virgin Cadmium-free Polypropylene