missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Microplates, 384-well
GREENER_CHOICE

Offers higher well density for economical use of expensive reagents. Fisherbrand™ Polypropylene Microplates, 384-well include round-shaped wells for optimal sample recovery.

Brand:  Fisherbrand™ 236200

 View more versions of this product

Product Code. 11977954

  • £366.00 / Pack of 120
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Ideal for storage of compounds and nucleic acid libraries
  • Chemical resistant
  • Withstands temperatures from -80° to +80°C
  • Conform to ANSI™ standard footprint for optimal use in automated handling and liquid-dispensing systems
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ANSI™ standard footprint
16 x 24
Polypropylene
Round
-80°C to +80°C
Standard
Yellow
384
120/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.