missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mutexil™ Visitor Protective Lab Coats
75.00 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Closure Type:
Snaps
Size:
X-Large
2X-Large
Description
PP non woven labcoat with press-stud fastening and pockets
- Disposable
- PP, 40g/m2
- For maintenance, production site visits
- For non hazardous environments
Specifications
Product Type | Lab Coat |
Size | X-Large |
Color | White |
Closure Type | Snaps |
No. of Pockets | None |
Packaging Type | Individually bagged |
Construction | Non-woven |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'