missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ PCR Tubes - Singles and Strips
GREENER_CHOICE

Product Code. 11843432 Shop All Eppendorf Products
Click to view available options
Closure Type:
Domed Strip Cap
Flat Strip Cap
Hinged Cap
Not Included
Format:
8-tube Strips
Fast PCR Tube Strips
Single Tube
Purity or Quality Grade:
Forensic DNA Grade
PCR Clean (Non-sterile)
Volume (Metric):
0.1 mL
0.2 mL
0.5 mL
Unit Size:
Pack of 1000
Pack of 120
Pack of 500
This item is not returnable. View return policy

Product Code. 11843432

Brand: Eppendorf™ 0030124804

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Eppendorf PCR Tubes ensure uniform heat distribution to the sample for optimal PCR performance. The user-friendly design allows for easy opening and provides a secure seal to prevent evaporation.

Ensure efficient and homogenous heat transfer with these thin-walled polypropylene tubes. Eppendorf™ Polypropylene PCR Tubes are easy to open, but provide tight sealing to prevent evaporation in PCR.

  • Design ensures an airtight seal to prevent contamination and evaboration.
  • Thin polypropylene walls ensure efficient and homogeneous heat transfer or fast and high-quality PCR results.
  • 8 reaction tubes in strip-format – ideal for small sample volumes. Easily adaptable for automation.
  • Strips are sealable using flat or domed strip-lids.
  • PCR Clean: Certified free from human DNA, DNase, RNase and PCR inhibitors).
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Human DNA, DNase, RNase, PCR inhibitor-free
Color Transparent
Volume (Metric) 0.1 mL
Quantity 120 Pk.
For Use With (Equipment) 0.1mL thermal cycler blocks for efficient and homogenous heat transfer to the sample
Sterility Non-sterile
Material Polypropylene
Purity or Quality Grade PCR Clean (Non-sterile)
Max. Force 10,000 x g
Closure Type Not Included
Autoclavable Autoclavable
Closure Included No
Format 8-tube Strips
Product Type PCR Tubes (8-tube Strip)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.