missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Polypropylene Photographic Trays
White stackable polypropylene (PP) photographic trays with ribbed bottom that improves stability.
Brand: BRAND™ 156642
This item is not returnable.
View return policy
Description
- White stackable polypropylene (PP) photographic trays
- Ribbed bottom improves stability, reinforced rim, slightly sloping walls
- Autoclavable (121 °C)
Specifications
300 x 240 x 70 mm | |
70 mm | |
Polypropylene | |
240 mm | |
Rectangular |
White | |
300 mm | |
Photographic Tray | |
1 | |
Autoclavable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction