Learn More
BRAND™ Polypropylene Photographic Trays
White stackable polypropylene (PP) photographic trays with ribbed bottom that improves stability.
Brand:  BRAND™ 156650
 View more versions of this product
Product Code. 10706711
  • £101.00 / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • White stackable polypropylene (PP) photographic trays 
  • Ribbed bottom improves stability, reinforced rim, slightly sloping walls
  • Autoclavable (121 °C)
  • TRUSTED_SUSTAINABILITY
Specifications
Specifications
160 mm