missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ Polypropylene Pipet Tip xl

£19.15 - £23.65

Specifications

Color Crystal
Autoclavable Autoclavable
Certifications/Compliance CE, IVD
For Use With (Application) Ideal for All Commercially Available Pippets
Sterilization Method Autoclavable
View More Specs

Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
10754794
View Documents Promotion Details
Ratiolab™
2100500
0.1 to 10 μL
£23.65
Pack of 1000
Estimated Shipment: 16-12-2024
to see stock.
Add to basket
 
10750363
View Documents Promotion Details
Ratiolab™
2500170
0.5 to 20 μL
£19.15
Pack of 1000
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

Crystal
CE, IVD
Autoclavable
Polypropylene
Autoclavable
Ideal for All Commercially Available Pippets
Universal Fit
Videos
SDS
Documents

Documents

Product Certifications