missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Polypropylene Plunger

Brand:  Cytiva 18-1031-81

37.60 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Code : GE

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 10327889

  • £40.00 / Each
Estimated Shipment: 06-08-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

XK 26, AK 26, and AK 16
Plunger
1 Pack
Polypropylene (Reinforced Glassfibre)
60°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers