missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Polypropylene Powder Funnels

Constructed of durable, lightweight polypropylene. Brand™ Polypropylene Powder Funnels are an excellent option for the transfer of samples without flow rate restrictions.

Brand:  BRAND™ 148045

9.03 GBP valid until 2024-11-29
Use promo code "21718" to get your promotional price.


 View more versions of this product

Product Code. 10777591

  • £16.05 / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket

Description

Description

  • With short, wide stem
  • Provides excellent resistance against thermal shock

TRUSTED_SUSTAINABILITY
Specifications

Specifications

180 mm
Polypropylene
Autoclavable
Filling Funnel (Powders)
40 mm
48 mm (Stem)
Translucent
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers