missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Polypropylene Powder Funnels

Constructed of durable, lightweight polypropylene. Brand™ Polypropylene Powder Funnels are an excellent option for the transfer of samples without flow rate restrictions.

Brand:  BRAND™ 148020

 View more versions of this product

Product Code. 11623139

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • With short, wide stem
  • Provides excellent resistance against thermal shock

TRUSTED_SUSTAINABILITY
Specifications

Specifications

65 mm
Polypropylene
Autoclavable
26 mm
Powder funnel
18 mm
26 mm (Stem)
Translucent
20/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers