Learn More
Cole-Parmer™ EPA Pre-Cleaned TOC Vial
Use these vials for cleanroom applications. Cole-Parmer™ EPA Pre-Cleaned TOC Vials are manufactured from clear borosilicate class and include attached white polypropylene hole cap and PTFE/silicone septa.
Brand: Cole-Parmer™ 03756-20
475.57 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Certified and traceable as associated with “Good Manufacturing Practices” (GMP)
- Includes Certificate of Analysis
- Packaged in an anti-static c-slotted plastic case
- Cleaned and certified for Total Organic Carbon (TOC) to <10ppb.
Specifications
Borosilicate Glass | |
40 mL | |
80 Pack |