Learn More
Brady™ Polypropylene Prinzing Ball Valve Lockout
The two halves of the Polypropylene Prinzing Ball Valve Lockout encompass the ball valve lever to secure it from inadvertent activation of the valve.
Brand:  Brady™ 805850
 View more versions of this product
Product Code. 11766785
  • £85.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The two halves of the device encompass the ball valve lever to secure it from inadvertent activation of the valve.
  • Locks out pipes from 12.70 to 31.75 mm in open and 12.70 to 63.50 mm closed position.
Specifications
Specifications
Valve Lockout
Blue
OSHA 1910.147
118.11 x 304.80 x 219.96 mm
Lockout Tagout
219.96 mm
12 in.
BS08A-BL
Polypropylene
Prinzing Ball Valve Lockout
305 x 220 mm
8.66 in.
1 Ea.
304.8 mm