missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Polypropylene Racks for Autosampler Vials

Product Code. 12672495 Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W x H):
200 x 105 x 17mm
230 x 117 x 28mm
For Use With (Equipment):
1.5mL vials
4mL Vials
Unit Size:
Each

Product Code. 12672495

Only null left
Add to basket
Add to basket

Polypropylene vial racks are resistant to most solvents.

  • Vial-Rack
  • 50 positions
  • Blue
  • Stackable
  • Feature alphanumeric indexing for easier vial identification
  • Stackable for efficient storage
TRUSTED_SUSTAINABILITY

Specifications

Product Type Vial Rack
Closure Included No
Autoclavable Autoclavable
Color Blue
Material Polypropylene
Positions 50
For Use With (Equipment) 4mL Vials
No. per Pack 1
Dimensions (L x W x H) 230 x 117 x 28mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.