missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ Polypropylene Reagent Bottle
Klik for at se tilgængelige muligheder
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
Closure Type:
Screw Cap with GL Thread
Stopper with Hexagonal Cap
Thread Size (Metric):
GL 25
GL 32
GL 63
NS
Pakningsstørrelse:
Pack of 10
Pack of 20
Pack of 6
Beskrivelse
Economical and durable plastic reagent bottles are ideal of storing liquids, powders and paste-like materials. A variety of sizes, seals (stopper or screw caps) and neck sizes are available to meet the storage needs of any lab material.
- Transparent media bottles ideal for long-term storage of chemicals
- Reagent bottles featuring screw caps or neck stoppers
- Stoppers: neck-size with a hexagonal-knob cap and red core
- Screw caps: high-pitch GL threads for significant carrying power
- Larger sizes (5 and 10L) available with handles for easier carrying
- Wider mouth bottles are ideal for easier filling, especially of powders and paste-like materials
- Constructed of study polypropylene
- Good chemical and temperature resistance
- Autoclavable at 121 °C (2 bar) according to DIN EN 285
Tekniske data
Tekniske data
| Capacity (Metric) | 2000 mL |
| For Use With (Application) | Long-term storage of liquids |
| Includes | Screw Cap |
| Material | Polypropylene |
| No. per Pack | 6 Bottles |
| Thread Size (Metric) | GL 63 |
| Mouth | Wide |
| Height (Metric) | 243 mm |
| Product Type | Reagent Bottle |
| Diameter (Metric) | 131 mm |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion