missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ Polypropylene Reagent Bottle
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
Closure Type:
Screw Cap with GL Thread
Stopper with Hexagonal Cap
Thread Size (Metric):
GL 25
GL 32
GL 63
NS
Unit Size:
Pack of 10
Pack of 20
Pack of 6
Description
Economical and durable plastic reagent bottles are ideal of storing liquids, powders and paste-like materials. A variety of sizes, seals (stopper or screw caps) and neck sizes are available to meet the storage needs of any lab material.
- Transparent media bottles ideal for long-term storage of chemicals
- Reagent bottles featuring screw caps or neck stoppers
- Stoppers: neck-size with a hexagonal-knob cap and red core
- Screw caps: high-pitch GL threads for significant carrying power
- Larger sizes (5 and 10L) available with handles for easier carrying
- Wider mouth bottles are ideal for easier filling, especially of powders and paste-like materials
- Constructed of study polypropylene
- Good chemical and temperature resistance
- Autoclavable at 121 °C (2 bar) according to DIN EN 285
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| For Use With (Application) | Long-term storage of liquids |
| Includes | Screw Cap |
| Material | Polypropylene |
| No. per Pack | 10 Bottles |
| Thread Size (Metric) | GL 25 |
| Height (Metric) | 165 mm |
| Product Type | Reagent Bottle |
| Diameter (Metric) | 87 mm |
| Closure Type | Screw Cap with GL Thread |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction