missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ Polypropylene Reagent Bottle

Product Code. 15291169
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
Closure Type:
Screw Cap with GL Thread
Stopper with Hexagonal Cap
Thread Size (Metric):
GL 25
GL 32
GL 63
NS
Unit Size:
Pack of 10
Pack of 20
Pack of 6
This item is not returnable. View return policy

Product Code. 15291169

Brand: VITLAB™ 101589

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Store liquid and powder chemicals securely. VITLAB™ Polypropylene Reagent Bottle is constructed of transparent, durable polypropylene for long-term storage. Choose from neck-size stoppers or maximum grip screw caps, both available in a variety of sizes.

Economical and durable plastic reagent bottles are ideal of storing liquids, powders and paste-like materials. A variety of sizes, seals (stopper or screw caps) and neck sizes are available to meet the storage needs of any lab material.
  • Transparent media bottles ideal for long-term storage of chemicals
  • Reagent bottles featuring screw caps or neck stoppers
    • Stoppers: neck-size with a hexagonal-knob cap and red core
    • Screw caps: high-pitch GL threads for significant carrying power
  • Larger sizes (5 and 10L) available with handles for easier carrying
  • Wider mouth bottles are ideal for easier filling, especially of powders and paste-like materials
  • Constructed of study polypropylene
    • Good chemical and temperature resistance
    • Autoclavable at 121 °C (2 bar) according to DIN EN 285

Specifications

Capacity (Metric) 100 mL
For Use With (Application) Long-term storage of liquids
Includes Screw Cap
Material Polypropylene
No. per Pack 20 Bottles
Thread Size (Metric) GL 32
Height (Metric) 96 mm
Product Type Reagent Bottle
Diameter (Metric) 55 mm
Closure Type Screw Cap with GL Thread
Autoclavable Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.