Learn More
VITLAB™ Polypropylene Reagent Bottle
Store liquid and powder chemicals securely. VITLAB™ Polypropylene Reagent Bottle is constructed of transparent, durable polypropylene for long-term storage. Choose from neck-size stoppers or maximum grip screw caps, both available in a variety of sizes.
Brand: VITLAB™ 101594
57.67 GBP valid until 2024-12-31
Use promo code "23277" to get your promotional price.
Additional Details : Weight : 1.67000kg
Description
Economical and durable plastic reagent bottles are ideal of storing liquids, powders and paste-like materials. A variety of sizes, seals (stopper or screw caps) and neck sizes are available to meet the storage needs of any lab material.- Transparent media bottles ideal for long-term storage of chemicals
- Reagent bottles featuring screw caps or neck stoppers
- Stoppers: neck-size with a hexagonal-knob cap and red core
- Screw caps: high-pitch GL threads for significant carrying power
- Larger sizes (5 and 10L) available with handles for easier carrying
- Wider mouth bottles are ideal for easier filling, especially of powders and paste-like materials
- Constructed of study polypropylene
- Good chemical and temperature resistance
- Autoclavable at 121 °C (2 bar) according to DIN EN 285
Specifications
100 mL | |
NS stopper made of Polypropylene and conical shoulders | |
20 Bottles | |
Autoclavable | |
111 mm | |
55 mm |
Long-term storage of liquids | |
Polypropylene | |
29/32 | |
Wide | |
Reagent Bottle | |
Stopper with Hexagonal Cap |