missing translation for 'onlineSavingsMsg'
Learn More

Vitlab™ Polypropylene Reagent Bottles

Use these bottles for long-term storage of liquids. VITLAB™ Polypropylene Reagent Bottles feature a wider mouth for simple filling of powders and paste-like materials.

£77.55

Specifications

Color Transparent
For Use With (Application) Ideal for powders and paste-like materials
Includes Screw cap
Material Polypropylene
Closure Included Yes
View More Specs

Products 1
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15201179
View Documents
Vitlab™
101889
1000 mL
£77.55
Pack of 10
Estimated Shipment: 19-12-2024
to see stock.
Add to basket
 
Description

Description

  • Transparent Reagent Bottles
  • Good chemical resistance
  • With polypropylene screw cap
  • Autoclavable at 121°C (2 bar) according to DIN EN 285
  • 5000mL size features one handle for easy transport and pouring
  • Suitable for foodstuffs
Specifications

Specifications

Transparent
Screw cap
Yes
Wide
Polypropylene
Ideal for powders and paste-like materials
Polypropylene
Autoclavable
Bottle
Screw Cap
Videos
SDS
Documents

Documents

Product Certifications