missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ PPCO Rectangular Carboy with Spigot

Click to view available options
Capacity (Metric):
9 L
20 L
Description
Value your lab can rely on. Fisherbrand PPCO Rectangular Carboys have a space-saving rectangular design is ideal for storing and dispensing lab water and reagents. Built to last. Made from high quality plastic materials. Autoclavable.
Ideal for storing and dispensing lab water and reagents
- Durable construction for long service life
- Molded graduations for 1 gallon and 51 increments
- Choice of materials for general use or autoclavability
- Space-saving rectangular design
- Wide mouth closure allows for easy filling and cleaning access
- Carboys with spigots include a hose barb insert to accommodate -+ to 5/16-in. ID tubing.
- Spigot spout without the adapter fits 5/8-in. ID tubing
- Materials meet FDA requirements for food use and USP VI criteria (non-toxic)
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy

Specifications
Specifications
Autoclavable | Autoclavable |
Capacity (English) | 2.4 gal. |
Capacity (Metric) | 9 L |
Certifications/Compliance | FDA, USP VI |
Closure Material | PPCO |
Color | White |
Dimensions (L x W x H) | External MaxD (with spigot) x W x H (with closure): 10.5 in. x 6.0 x 14.2 in. (26.7 x 15.3 x 36.1 cm) |
For Use With (Application) | Ideal for storing and dispensing lab water and reagents |
Material | PPCO |
Mouth Style | Wide |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Videos
-
Fisherbrand Rectangular Carboys (EN)
Documents
Scientific Resources
Brochures
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ PPCO Rectangular Carboy with Spigot >
Spot an opportunity for improvement?Share a Content Correction