missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Polypropylene, Rectangular Carboy with Spigot
61.53 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Use promo code "23291" to get your promotional price.
Click to view available options
Capacity (Metric):
9 L
20 L
Description
Value your lab can rely on. Fisherbrand Polypropylene Rectangular Carboys have a space-saving rectangular design is ideal for storing and dispensing lab water and reagents. Built to last. Made from high quality plastic materials. Autoclavable.
Ideal for storing and dispensing lab water and reagents
- Durable construction for long service life
- Molded graduations for 1 gallon and 51 increments
- Choice of materials for general use or autoclavability
- Space-saving rectangular design
- Wide mouth closure allows for easy filling and cleaning access
- Carboys with spigots include a hose barb insert to accommodate -+ to 5/16-in. ID tubing.
- Spigot spout without the adapter fits 5/8-in. ID tubing
- Materials meet FDA requirements for food use and USP VI criteria (non-toxic)
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Autoclavable | Autoclavable |
Capacity (English) | 5.3 gal. |
Capacity (Metric) | 20 L |
Certifications/Compliance | FDA, USP VI |
Closure Material | Polypropylene |
Color | White |
Dimensions (L x W x H) | External MaxD (with spigot) x W x H (with closure): 14.5 x 9.0 x 15.7 in. (36.1 x 22.9 x 39.9 cm) |
For Use With (Application) | Ideal for storing and dispensing lab water and reagents |
Material | Polypropylene |
Mouth Style | Wide |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'